Learn More
Honeywell IntelliDoX Docking Station
Honeywell IntelliDoX Docking Station is a bump test and calibration station for gas detectors that demonstrates quick and efficient performance.
Supplier: Honeywell DXBWC4
Description
Honeywell IntelliDoX Docking Station is a bump test and calibration station for gas detectors that demonstrates quick and efficient performance.
- High-tech way to combine smart docking modules and Fleet Manager II software to provide automated instrument management to drive productivity, reliability and efficiency.
- Enables bump test tracking, email notices and easy recordkeeping for improved compliance
- Performs a quick bump test for increased uptime and no wasted calibration gas
- Enables the ability to change alarm set points, set a noncompliance indicator when a bump test is due, and adjust other parameters for maximum flexibility
- IntelliDoX module comes with factory-installed nest
- Power cord, calibration gas tubing and exhaust tubing are packaged separately
- One Enabler Kit is required for each stand-alone IntelliDoX module
- 2-year warranty
Specifications
Certicate of Compliance; CSA/UL 61010-1 (3rd Ed); EC Declaration of Conformity; EMC Directive & Low Voltage Directive | |
BW Clip4 | |
13.8 cm | |
1.91 kg |
13.8 x 36.2 x 10.9 cm | |
Gas Detection Monitor Test Stations | |
36.2 cm | |
10.9 cm |