Learn More
Honeywell IntelliDoX Enabler Kit, (North American Version)
Catalog No. 19807576
Click to view available options
For Use With (Equipment):
IntelliDoX Docking Module
Catalog No. For Use With (Equipment)
19-807-576 IntelliDoX Docking Module
1 options
Catalog No. 19-807-576 Supplier Honeywell Supplier No. DXENBLNA
IntelliDoX Enabler Kit combines smart docking modules and Fleet Manager II software to provide automated instrument management to help drive productivity, reliability and efficiency.
  • Enables bump test tracking, email notices and easy recordkeeping for improved compliance
  • Performs the quick bump test for increased uptime and no wasted calibration gas
  • Enables the ability to change alarm set points, set a noncompliance indicator when a bump test is due, and adjust other parameters for maximum flexibility
  • 2-year warranty
  • Includes IntelliDoX module with a factory-installed nest
  • Power adapter, power cord, calibration gas tubing and exhaust tubing, are packaged
    separately
  • One Enabler Kit is required for each stand-alone IntelliDoX module
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance Certicate of Compliance; CSA/UL 61010-1 (3rd Ed); EC Declaration of Conformity; EMC Directive & Low Voltage Directive
For Use With (Equipment) IntelliDoX Docking Module
Height (Metric) 13.8 cm
Length (Metric) 36.2 cm
Weight (Metric) 1.91 kg
Width (Metric) 10.9 cm