Learn More
GSC Go Science Crazy Intelligent Timner with Solenoid Power Supply
Description
The Intelligent Timer allows you to accurately record speeds of objects with consistent release times. This timer works with our entire line of solenoid and photogate products. With this timer you can control one solenoid's release and up to two photogates. The buttons on the front include the following: Start (to release the solenoid or start measuring with the photogates), Mode (between ldquo;Distancerdquo; and lqduo;Timerdquo;), Set Distance, Store Memory, Result, and Reset.
Specifications
Specifications
Product Type | Intelligent Timer |
For Use With (Application) | Accurately record speeds of objects with consistent release times |
For Use With (Equipment) | Solenoids and photogates |
Includes | Power cord |