Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Intercalated DNA Monoclonal Antibody (ET602.2), QED Bioscience™

Mouse Monoclonal Antibody
$250.22 - $1002.00
Specifications
Antigen | Intercalated DNA |
---|---|
Clone | ET602.2 |
Concentration | 0.5-1.0 mg/mL |
Applications | ELISA |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-227-7457
![]() ![]() |
Leinco Technologies Inc
12401100UG |
100 μg |
Each for $250.22
|
|
|||||
50-227-7458
![]() ![]() |
Leinco Technologies Inc
12401500UG |
500 μg |
Each for $500.43
|
|
|||||
50-227-7456
![]() ![]() |
Leinco Technologies Inc
124011000UG |
1 mg |
Each for $1,002.00
|
|
|||||
Description
Intercalated DNA Monoclonal antibody specifically detects Intercalated DNA in Chemical samples. It is validated for ELISASpecifications
Intercalated DNA | |
0.5-1.0 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
Ethidium bromide intercalated calf thymus DNA. | |
Primary | |
-20° C, Avoid Freeze/Thaw Cycles |
ET602.2 | |
ELISA | |
Unconjugated | |
Mouse | |
PBS with no preservative; pH 7.4 | |
IgG2b | |
Protein A | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title