Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Interferon alpha-10 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5116700
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5116700 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5116700 Supplier Invitrogen™ Supplier No. PA5116700
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive Samples: Rat plasma Immunogen sequence: CDLPQTHSLG NRRALILLGQ MGRISPFSCL KDRHDFRIPQ EEFDGNQFQK AQAISVLHEM IQQTFNLFST EDSSAAW.

Produced by macrophages, IFN-alpha have antiviral activities. Interferon stimulates the production of two enzymes: a protein kinase and an oligoadenylate synthetase.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Interferon alpha-10
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.67 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene IFNA10
Gene Accession No. P01566
Gene Alias IFN alpha; IFNA10; IFN-alpha-10; IFN-alphaC; Interferon; interferon alpha 10; interferon alpha-10; Interferon alpha-6L; interferon alpha-C; interferon, alpha 10; leIF C
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 24-100 of human IFNA10 (NP_0021621).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3446
Target Species Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.