Learn More
Invitrogen™ Interferon alpha-2 Polyclonal Antibody

Description
Recommended positive controls: A431, H1299, HeLa, HepG2. Predicted reactivity: Rhesus Monkey (100%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Specifications
Specifications
| Antigen | Interferon alpha-2 |
| Applications | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | 0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 |
| Gene | IFNA2 |
| Gene Accession No. | P01563 |
| Gene Alias | alpha-2a interferon; Ifa2; IFNA; IFN-a; IFNA 2; Ifna2; IFNA2A; IFNA2B; IFNA2C; IFN-alpha 1b; IFN-alpha-1/13; IFN-alpha-2; IFN-alphaA; INFA2; Interferon; interferon alpha 1b; interferon alpha 2; interferon alpha 2a; interferon alpha 2b; interferon alpha A; interferon alpha family, gene 2; interferon alpha-2; Interferon alpha-A; interferon alpha-D; interferon, alpha 2; Interferon-alpha II; leIF A; leIF D; RP11-354P17.2 |
| Gene Symbols | IFNA2 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.