Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Interferon alpha-2 Recombinant Rabbit Monoclonal Antibody (5P3U6), Invitrogen™

Catalog No. FEMA551384
Encompass
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEMA551384 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEMA551384 Supplier Thermo Scientific Supplier No. MA551384

Rabbit Recombinant Monoclonal Antibody

Immunogen sequence: CDLPQTHSLG SRRTLMLLAQ MRRISLFSCL KDRHDFGFPQ EEFGNQFQKA ETIPVLHEMI QQIFNLFSTK DSSAAWDETL LDKFYTELYQ QLNDLEACVI QGVGVTETPL MKEDSILAVR KYFQRITLYL KEKKYSPCAW EVVRAEIMRS FSLSTNLQES LRSKE

Interferon alpha-2 Recombinant Monoclonal antibody specifically detects Interferon alpha-2 in Human, Mouse, Rat samples. It is validated for Immunohistochemistry (Paraffin), Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen Interferon alpha-2
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Recombinant Monoclonal
Clone 5P3U6
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% BSA, 50% glycerol and 0.05% ProClin 300; pH 7.3
Gene IFNA2
Gene Accession No. P01563, P01573
Gene Alias alpha-2a interferon, Ifa2, IFNA, IFN-a, IFNA 2, Ifna2, IFNA2A, IFNA2B, IFNA2C, IFN-alpha 1b, IFN-alpha-1/13, IFN-alpha-2, IFN-alphaA, INFA2, interferon alpha 1b, interferon alpha 2, interferon alpha 2a, interferon alpha 2b, interferon alpha A, interferon alpha family, gene 2, interferon alpha-2, Interferon alpha-A, interferon alpha-D, interferon, alpha 2, Interferon-alpha II, leIF A, leIF D, RP11-354P17.2
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 24-188 of human Interferon alpha 2 (IFN-alpha 2) (NP_000596.2).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 15965, 298213, 3440
Target Species Human, Mouse, Rat
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Format Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.