Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Interferon alpha-21 Monoclonal Antibody (d4), Invitrogen™
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA543686
This item is not returnable.
View return policy
Description
Produced by macrophages, IFN-alpha have antiviral activities. Interferon stimulates the production of two enzymes: a protein kinase and an oligoadenylate synthetase.
Specifications
Interferon alpha-21 | |
Monoclonal | |
1 mg/mL | |
PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
P01568 | |
IFNA21 | |
Asp25-Glu189 (Accession P01568), with N-terminal His and GST Tag. | |
100 μL | |
Primary | |
Human, Pig | |
Antibody | |
IgG2b κ |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
d4 | |
Unconjugated | |
IFNA21 | |
IFNA21; IFNA5; IFNA6; IFN-alpha-21; IFN-alpha-5; IFN-alpha-6; IFN-alphaG; IFN-alphaI; IFN-alphaK; INA5; INFA5; Interferon; interferon alpha 21; interferon alpha 5; interferon alpha 6; interferon alpha-21; Interferon alpha-5; interferon alpha-54; Interferon alpha-6; interferon alpha-61; Interferon alpha-F; interferon alpha-G; Interferon alpha-K; interferon, alpha 21; interferon, alpha 5; interferon, alpha 6; LeIF F; LeIF G; LeIF K; leIF-F; leukocyte interferon protein | |
Mouse | |
Protein A/G | |
RUO | |
3452 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction