Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Interferon alpha-21 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA568034
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA568034 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA568034 Supplier Invitrogen™ Supplier No. PA568034
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Interferon alpha-21 Polyclonal Antibody for Western Blot

Produced by macrophages, IFN-alpha have antiviral activities. Interferon stimulates the production of two enzymes: a protein kinase and an oligoadenylate synthetase.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Interferon alpha-21
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene IFNA21
Gene Accession No. P01568
Gene Alias IFNA21; IFNA5; IFNA6; IFN-alpha-21; IFN-alpha-5; IFN-alpha-6; IFN-alphaG; IFN-alphaI; IFN-alphaK; INA5; INFA5; Interferon; interferon alpha 21; interferon alpha 5; interferon alpha 6; interferon alpha-21; Interferon alpha-5; interferon alpha-54; Interferon alpha-6; interferon alpha-61; Interferon alpha-F; interferon alpha-G; Interferon alpha-K; interferon, alpha 21; interferon, alpha 5; interferon, alpha 6; LeIF F; LeIF G; LeIF K; leIF-F; leukocyte interferon protein
Gene Symbols IFNA21
Host Species Rabbit
Immunogen Synthesized peptide derived from human Interferon alpha-21.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3452
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.