Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Interferon alpha-4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA535276
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA535276 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA535276 Supplier Invitrogen™ Supplier No. PA535276
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Interferon alpha-4 Polyclonal Antibody for Western Blot, IHC (P)

Produced by macrophages, IFN-alpha have antiviral activities. Interferon stimulates the production of two enzymes: a protein kinase and an oligoadenylate synthetase.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Interferon alpha-4
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene IFNA4
Gene Accession No. P05014
Gene Alias Ifa4; IFNA4; IFN-alpha-4; IFN-alpha4a; INFA4; Interferon; interferon alpha 4; interferon alpha family, gene 4; interferon alpha-4; Interferon alpha-4B; interferon alpha-76; Interferon alpha-M1; interferon, alpha 4; MuIFN-alpha 4
Gene Symbols IFNA4
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 162-189 amino acids from the C-terminal region of human IFNA4.
Purification Method Antigen affinity chromatography, Protein A
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3441
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.