Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Interferon alpha-4/10/14 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5100829
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5100829 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5100829 Supplier Invitrogen™ Supplier No. PA5100829
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total IFNA4/10/14.

IFNA4 (Interferon Alpha 4) and IFNA14 (Interferon Alpha 14) are predicted to enable cytokine activity and type I interferon receptor binding activity and to be involved in several processes, including B cell activation; lymphocyte activation involved in immune response; and positive regulation of peptidyl-serine phosphorylation of STAT protein. IFNA10 (Interferon Alpha 10) encodes a protein that belongs to the type I interferon family of proteins, and is located in a cluster of alpha interferon genes on chromosome 9. Interferons are small regulatory molecules that function in cell signaling in response to viruses and other pathogens or tumor cells. This gene is intronless and the encoded protein is secreted.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Interferon alpha-4/10/14
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene IFNA10
Gene Accession No. P01566, P01570, P05014, P07351
Gene Alias Interferon
Gene Symbols IFNA10, IFNA14, IFNA4
Host Species Rabbit
Immunogen A synthesized peptide derived from human IFNA4/10/14, corresponding to a region within C-terminal amino acids.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 110296, 15967, 3441, 3446, 3448, 404549
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.