Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Interferon alpha-6 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5103427
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5103427 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5103427 Supplier Invitrogen™ Supplier No. PA5103427
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total IFNA6.

Produced by macrophages, IFN-alpha have antiviral activities. Interferon stimulates the production of two enzymes: a protein kinase and an oligoadenylate synthetase.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Interferon alpha-6
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene IFNA6
Gene Accession No. P05013, P07350
Gene Alias IFNA6; IFN-alpha-6; IFN-alphaK; Interferon; interferon alpha 6; interferon alpha-54; Interferon alpha-6; interferon alpha-K; interferon, alpha 6; leIF K; RP11-354P17.7
Gene Symbols IFNA6
Host Species Rabbit
Immunogen A synthesized peptide derived from human IFNA6(Accession P05013), corresponding to amino acid residues T151-E189.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 15969, 3443
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.