Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Interleukin-1β, Rabbit, Unlabeled, Polyclonal,

Catalog No. AB1413IMI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
AB1413IMI 100 μg Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AB1413IMI Supplier MilliporeSigma™ Supplier No. AB1413I
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Interleukin-1 beta (UniProt; also known as IL-1 beta, IL-1b, IL-1beta) is encoded by the Il1b gene (Gene ID 16176) in murine species. Interleukin-1 (IL-1) is a cytokine that plays a central role in the regulation of acute and chronic inflammation. Two forms of IL-1, IL-1 and IL-1 , are transcribed from two separate genes and mediate similar, but not completely overlapping, biological effects via the membrane-bound IL-1 type 1 receptor (IL-1R1). Studies utilizing Il1a- and Il1b-knockout mice showed that IL-1 , but not IL-1 , is involved in T-cell-dependent antibody production via antigen-specific T-cell activation. IL-1 binding to the extracellular domain of IL1R1 promotes the recruitment of the IL-1 receptor accessory protein and subsequent signal transduction mediated by the cytoplasmic domains of IL-1R1 and IL1-R2. The IL-1 type 2 receptor (IL1R2) is unable to initiate signaling and only acts as a decoy receptor. Soluble receptors for IL-1 (sIL-1Rs) are produced by proteolytic cleavage of membrane-bound receptors or by alternative splicing and act as natural inhibitors by competing against membrane-bound IL1Rs for IL-1 binding. Murine IL-1 beta is produced with a propeptide sequence (a.a. 1-117), which is removed posttranslationally to yield the mature (a.a. 118-269) protein.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Interleukin-1β
Applications Western Blot
Classification Polyclonal
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Formulation Purified rabbit polyclonal antibody in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% Sodium Azide.
Gene Il1b
Gene Accession No. P10749
Host Species Rabbit
Immunogen KLH-conjugated linear peptide corresponding to a region within the C-terminal half of the mature murine and rat IL-1beta sequence.
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Inflammation & Immunology
Primary or Secondary Primary
Gene ID (Entrez) NP_032387
Test Specificity This rabbit polyclonal antibody detected both the ∽17 kDa mature and ∽30 kDa pro-form of murine IL-1beta. The corresponding sequence in human IL-1beta differs from that of the immunogen by only one amino acid. Immunogen sequence is not found in IL-1alpha.
Target Species Mouse
Content And Storage Stable for 1 year at 2°-8°C from date of receipt.
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.