Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

International Enviroguard™ MicroGuard CE™ Clean Processed Microporous Boot Covers

Catalog No. p-4162784 Shop All International Enviroguard Systems Products
Click to view available options
Size:
Large
X-Large
Thickness:
25 mil
26 mil
27 mil
28 mil
29 mil
30 mil
Packaging Type:
Bulk Packaged
Individually Packaged
This item is not returnable. View return policy

Catalog No. 19-472-558

Supplier: International Enviroguard™ CE8104CIL

Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Disposable boot covers are made from high MVTR, innovative fabric. Ideal for ISO 5 or higher non-hazardous work environments where reliable liquid and particulate protection is needed.

These microporous boot covers are clean processed to offer the perfect balance of safety and comfort. The fabric keeps dust and liquids out while allowing air to flow freely, so your feet stay cool throughout a hard day's work. These disposable boot covers are ideal for non-hazardous work environments, such as controlled environments or cleanrooms, where you need reliable liquid and particulate protection. Boot covers are designed with tunnelized elastic to contain particle shed. Non-skid soles provide added traction to help prevent accidental slips and trips. Each boot cover is individually packaged to help prevent cross-contamination.

  • Non-skid soles help improve traction to prevent accidental slips
  • Elastic closures for snug, secure fit that keeps boot covers in place
  • Available in Bulk or Individually packaged to prevent cross-contamination

Specifications

Size Large
Material Microporous Fabric
Color White
Closure Type Elastic Top and Ankles
Sole Type Non-skid
Seam Type Serged
Thickness 28 mil
Certifications/Compliance ANSI ISEA 101-1996 R2020, ASTM T903, ASTM F1670-07
Packaging Quantity 200/Cs.
Packaging Type Individually Packaged
Height (English) 17 in.
Height (Metric) 43.18 cm
Product Type Controlled Environments Boot Covers
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.