Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
International Enviroguard™ MicroGuard CE™ Cleanroom Frocks
Shop All International Enviroguard Systems Products
Click to view available options
Size:
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
Length (English):
43 in.
44 in.
46 in.
47 in.
48.5 in.
50 in.
Size (English) Chest:
38 in.
40 in.
42 in.
44 in.
45 in.
46 in.
Packaging Type:
Bulk Packaged
Individually Packaged
Description
- Microporous, clean processed frocks available in individual or bulk packaging
- Fabric keeps dust and liquids out while allowing air to flow freely
- Ideal for non-hazardous work environments where reliable liquid and particulate protection is needed
- Designed with tunnelized elastic to contain particle shed
- Bound mandarin collar for enhanced protection
- Front snap closures are easy to secure
- Constructed with bound seams to eliminate raw edges and strengthen the seam
- No pockets
Specifications
Specifications
| Product Type | Controlled Environments Frock |
| Size | X-Large |
| Material | Microporous Fabric |
| Color | White |
| Disposable | Single-use |
| Sterility | Non-sterile |
| Closure Type | Snaps |
| Cuff Style | Elastic |
| Seam Type | Bound |
| Collar Type | Bound Mandarin |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction