Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

International Enviroguard™ MicroGuard CE™ Cleanroom Frocks

Catalog No. 19472546
Click to view available options
Size:
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
Length (English):
43 in.
44 in.
46 in.
47 in.
48.5 in.
50 in.
Size (English) Chest:
38 in.
40 in.
42 in.
44 in.
45 in.
46 in.
Packaging Type:
Bulk Packaged
Individually Packaged
This item is not returnable. View return policy
This item is not returnable. View return policy

Cleanroom frocks utilize state-of-the-art lamination technology to combine a soft non-woven fabric with a microporous film.

  • Microporous, clean processed frocks available in individual or bulk packaging
  • Fabric keeps dust and liquids out while allowing air to flow freely
  • Ideal for non-hazardous work environments where reliable liquid and particulate protection is needed
  • Designed with tunnelized elastic to contain particle shed
  • Bound mandarin collar for enhanced protection
  • Front snap closures are easy to secure
  • Constructed with bound seams to eliminate raw edges and strengthen the seam
  • No pockets

Specifications

Product Type Controlled Environments Frock
Size 2X-Large
Material Microporous Fabric
Color White
Disposable Single-use
Sterility Non-sterile
Closure Type Snaps
Cuff Style Elastic
Seam Type Bound
Collar Type Bound Mandarin
Size (English) Chest 44 in.
Size (Metric) Chest 111.7 cm
Certifications/Compliance CE
Packaging Type Individually Packaged
Length (English) 47 in.
Length (Metric) 119.3 cm
Weight (English) 2.1 oz.
Weight (Metric) 0.05 kg
Cleanroom Class Class 100 (ISO 5)
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.