Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
International Enviroguard™ MicroGuard CE™ Clean Processed Microporous Shoe Covers
Click to view available options
Size:
Large
X-Large
Thickness:
19 mil
20 mil
21 mil
22 mil
23 mil
24 mil
Packaging Type:
Bulk Packaged
Individually Packaged
Description
- Clean-processed shoe covers provide protection against particulates and non-hazardous liquids
- Breathable fabric allows moisture vapor to escape to help keep feet cooler
- Tunnelized elastic at the top contains particle shed and eliminates raw edges
- Environmentally friendly and can be recycled after use
- Non-skid soles help improve traction to prevent accidental slips
- Elastic closures for snug, secure fit that keeps boot covers in place
- Individually packaged
Specifications
Specifications
Size | X-Large |
Material | Microporous Fabric |
Color | White |
Sterility | Non-sterile |
Closure Type | Elastic Ankle |
Sole Type | Non-skid |
Seam Type | Serged |
Thickness | 24 mil |
Certifications/Compliance | ANSI ISEA 101-1996 R2020, ASTM T903, ASTM 1670-07 |
Packaging Quantity | 200/Cs. |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction