Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

International Enviroguard™ PVC Cleanroom Chemical Aprons with Long Sleeves

Catalog No. 19472672 Shop All International Enviroguard Systems Products
Click to view available options
Size:
Medium
X-Large
2X-Large
This item is not returnable. View return policy
This item is not returnable. View return policy

Heavy-duty PVC aprons features long sleeves with tunnelized elastic wrists for protection against liquid splash, sprays, and non-hazardous dry particulates.

  • Heavy-duty, 6 mil thick chemical-resistant aprons are made of PVC
  • Designed to protect against acids, alcohol, oils, grease, fats, and a variety of chemicals
  • Liquid-resistant, abrasion-resistant, and puncture-resistant to withstand tough work environments
  • Protect against liquid splash, sprays, and non-hazardous dry particulates
  • Loop over the head and nylon ties on the back
  • Edges are ultrasonically welded to reduce linting and release of unwanted particulates
  • Offer full protection of the front of the body from the chest to the knees or below
  • Long sleeves with tunnelized elastic cuffs to protect the arms from splashes and sprays and to also increase particle containment
  • Cleanroom-ready aprons are clean-processed and individually bagged
  • Applications include semi-conductor, cleanroom, pharmaceutical, chemical processing, petrochemicals, sanitation work
  • Latex-free elastic

Specifications

Type Controlled Environments Apron
Size X-Large
Material PVC
Color Blue
Disposable Single-use
Closure Type Waist Ties
Sterility Non-sterile
Resistance Liquid, Abrasion, Puncture
Thickness 6 mil
For Use With (Application) Semi-conductor, Cleanroom, Pharmaceutical, Chemical Processing, Petrochemicals, Sanitation Work
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.