Learn More
International Enviroguard™ PVC Chemical Apron with Long Sleeves
Catalog No. 19472676 Shop All International Enviroguard Systems Products
Click to view available options
Size:
Medium
Large
X-Large
2X-Large
This item is not returnable. View return policy
Catalog No. 19-472-676
Supplier: International Enviroguard™ IT6CABLC
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy
Heavy-duty, Blue PVC Chemical Apron with Long Sleeves for protection against acids, chemicals, alcohol, oils, grease, and more.
These heavy-duty, chemical-resistant aprons are made of PVC and designed to protect against acids, alcohol, oils, grease, fats, and a variety of chemicals. PVC aprons are liquid-resistant, abrasion-resistant, and puncture-resistant to withstand tough work environments. Aprons also protect against liquid splash, sprays, and non-hazardous dry particulates. Each apron has a loop to go over the head and nylon ties on the back. All edges are ultrasonically welded to reduce linting and the release of unwanted particulates into the cleanroom environment.
Aprons offer full protection of the front of the body from the chest to the knees or below. These PVC-coated aprons have long sleeves with tunnelized elastic cuffs to protect the arms from splashes and sprays and to also increase particle containment. Cleanroom-ready aprons are clean-processed and individually bagged under cleanroom conditions for use in the most critical controlled environments. These chemically coated aprons are versatile with possible applications including semi-conductor, cleanroom, pharmaceutical, chemical processing, petrochemicals, sanitation work, and more.
  • 6 mil thick PVC-coated, long-sleeved apron protects against chemicals, non-hazardous particulates, acids, oils, splashes, sprays, and more
  • Made with latex-free elastic; ties at the neck and waist in the back
  • Resists penetration of various workplace chemicals such as acetone, sodium hydroxide, nitric acid, isopropyl alcohol, and more
  • Protects the full front side of the body from the chest to the knees or below
  • Double-sealed, tunnelized elastic wrists contain particle shed
Specifications
Type Apron
Size Large
Material PVC
Color Blue
Disposable Single-use
Closure Type Waist Ties
Cuff Style Elastic
Resistance Liquid, Abrasion, and Puncture Resistant
Thickness 6 mil
No. per Case 36/Cs.
Seam Type Welded
Show More Show Less
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.