Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Intersectin 1 Antibody, Alexa Fluor™ 532, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP171832AF532
Description
Intersectin 1 Polyclonal antibody specifically detects Intersectin 1 in Human, Mouse samples. It is validated for Western BlotSpecifications
Intersectin 1 | |
Polyclonal | |
Western Blot | |
11, intersectin 1 (SH3 domain protein), intersectin short variant 12, intersectin-1, ITSN, SH3 domain protein-1A, SH3 domain-containing protein 1A, Src homology 3 domain-containing protein | |
The immunogen for this product maps to a region between residue 850 and 900 of human Intersectin 1 using the numbering given in entry NP_003015.2 (GeneID 6453). | |
0.1 mL | |
Primary | |
Human, Mouse | |
Purified |
Western Blot | |
Alexa Fluor 532 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
6453 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction