Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Intersectin 1 Antibody, DyLight 650, Novus Biologicals™
SDP

Catalog No. NB262849 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB262849 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB262849 Supplier Novus Biologicals Supplier No. NBP171832C
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Intersectin 1 Polyclonal antibody specifically detects Intersectin 1 in Human, Mouse samples. It is validated for Western Blot

Specifications

Antigen Intersectin 1
Applications Western Blot
Classification Polyclonal
Conjugate DyLight 650
Dilution Western Blot
Formulation 50mM Sodium Borate
Gene Alias 11, intersectin 1 (SH3 domain protein), intersectin short variant 12, intersectin-1, ITSN, SH3 domain protein-1A, SH3 domain-containing protein 1A, Src homology 3 domain-containing protein
Host Species Rabbit
Immunogen The immunogen for this product maps to a region between residue 850 and 900 of human Intersectin 1 using the numbering given in entry NP_003015.2 (GeneID 6453).
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6453
Target Species Human, Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.