Learn More
Intersectin 1 Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A303075AT

Description
The recommended shelf life for this product is 1 year from date of receipt.
Adapter protein that may provide indirect link between the endocytic membrane traffic and the actin assembly machinery. May regulate the formation of clathrin-coated vesicles. Isoform 1 could be involved in brain-specific synaptic vesicle recycling.Specifications
Intersectin 1 | |
Polyclonal | |
TBS with 0.1% BSA and 0.09% sodium azide | |
Q15811 | |
ITSN1 | |
Between 850 and 900 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Western Blot | |
Unconjugated | |
ITSN1 | |
ITSN, MGC134948, MGC134949, SH3D1A, SH3P17 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
6453 | |
4° C | |
Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.