Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Intestinal Alkaline Phosphatase Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA596536
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA596536 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA596536 Supplier Invitrogen™ Supplier No. PA596536
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive Samples: HepG2; Cellular Location: Cell membrane, GPI-anchor, Lipid-anchor Immunogen sequence: VIPAEEENPA FWNRQAAEAL DAAKKLQPIQ KVAKNLILFL GDGLGVPTVT ATRILKGQKN GKLGPETPLA MDRFPYLALS KTYNVDRQVP DSAATATAYL CGVKANFQTI GLSAAARFNQ CNTTRGNEVI SVMNRAKQAG KSVGVVTTTR VQHASPAGTY AHTVNRNWYS DADMPASARQ E.

There are at least four distinct but related alkaline phosphatases: intestinal, placental, placental-like, and liver/bone/kidney (tissue non-specific). The intestinal alkaline phosphatase gene encodes a digestive brush-border enzyme. This enzyme is upregulated during small intestinal epithelial cell differentiation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Intestinal Alkaline Phosphatase
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.34 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene ALPI
Gene Accession No. P09923
Gene Alias 2010001C14Rik; Akp2; Akp6; Alkaline phosphatase 1, intestinal, defined by SSR; alkaline phosphatase 6; alkaline phosphatase, intestinal; alkaline phosphomonoesterase; Alp1; ALPI; AP; EC 3.1.3.1; gIAP; glycerophosphatase; I Kasahara isozyme; IAP; IAP-1; IAP-I; intestinal alkaline phosphatase; intestinal alkaline phosphatase 1; intestinal alkaline phosphatase I IAP-I; intestinal alkaline phosphatase III; intestinal alkaline phosphatase IV; intestinal alkaline phosphatase V; intestinal-type alkaline phosphatase; intestinal-type alkaline phosphatase 1; Kasahara isozyme; PLAP-1; Regan isozyme; S51097
Gene Symbols ALPI
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 20-200 of human ALPI (NP_0016222).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 248
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.