Learn More
Invitrogen™ Intestinal Alkaline Phosphatase Recombinant Rabbit Monoclonal Antibody (001)
Rabbit Recombinant Monoclonal Antibody
Supplier: Invitrogen™ MA530076
Description
This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). Recombinant rabbit monoclonal antibodies are produced using in vitro expression systems. The expression systems are developed by cloning in the specific antibody DNA sequences from immunoreactive rabbits. Then, individual clones are screened to select the best candidates for production. The advantages of using recombinant rabbit monoclonal antibodies include: better specificity and sensitivity, lot-to-lot consistency, animal origin-free formulations, and broader immunoreactivity to diverse targets due to larger rabbit immune repertoire. This antibody has specificity for Human ALPI.
There are at least four distinct but related alkaline phosphatases: intestinal, placental, placental-like, and liver/bone/kidney (tissue non-specific). The intestinal alkaline phosphatase gene encodes a digestive brush-border enzyme. This enzyme is upregulated during small intestinal epithelial cell differentiation.
Specifications
Intestinal Alkaline Phosphatase | |
Recombinant Monoclonal | |
1 mg/mL | |
PBS with no preservative | |
P09923 | |
ALPI | |
Recombinant Human ALPI protein (Met1-Asp503). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
ELISA | |
001 | |
Unconjugated | |
ALPI | |
2010001C14Rik; Akp2; Akp6; Alkaline phosphatase 1, intestinal, defined by SSR; alkaline phosphatase 6; alkaline phosphatase, intestinal; alkaline phosphomonoesterase; Alp1; ALPI; AP; EC 3.1.3.1; gIAP; glycerophosphatase; I Kasahara isozyme; IAP; IAP-1; IAP-I; intestinal alkaline phosphatase; intestinal alkaline phosphatase 1; intestinal alkaline phosphatase I IAP-I; intestinal alkaline phosphatase III; intestinal alkaline phosphatase IV; intestinal alkaline phosphatase V; intestinal-type alkaline phosphatase; intestinal-type alkaline phosphatase 1; Kasahara isozyme; PLAP-1; Regan isozyme; S51097 | |
Rabbit | |
Protein A | |
RUO | |
248 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.