Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Intra Acrosomal Protein Antibody (Ds-2), DyLight 755, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB500436IR
Description
Intra Acrosomal Protein Monoclonal specifically detects Intra Acrosomal Protein in Canine samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Specifications
Intra Acrosomal Protein | |
Monoclonal | |
DyLight 755 | |
acrosomal protein SP-10, acrosomal vesicle protein 1D11S4365, SP-10, SPACA2, sperm protein 10 | |
Mouse | |
Affinity Purified | |
RUO | |
56 | |
Canine | |
IgG |
Western Blot, Immunocytochemistry, Immunofluorescence | |
Ds-2 | |
Western Blot, Immunocytochemistry/Immunofluorescence | |
ACRV1 | |
Freshly ejaculated dog sperms were washed in PBS and extracted in 3% acetic acid, 10% glycerol, 30 mM benzaminidine. The acid extract was dialyzed against 0.2% acetic acid, lyophylized and subsequently used for immunization. | |
0.1 mL | |
Primary | |
The antibody Ds-2 reacts with canine intra-acrosomal sperm protein (92 and 42 kDa), a testis-specific acrosomal vesicle protein 1 associated with the acrosomal membranes and matrix of the mature sperm. | |
Store at 4C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction