Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Intra Acrosomal Protein Antibody (Ds-2), Janelia Fluor™ 646, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB500436JF646
Description
Intra Acrosomal Protein Monoclonal specifically detects Intra Acrosomal Protein in Canine samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Specifications
IntraAcrosomalProtein | |
Monoclonal | |
Janelia Fluor 646 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
ACRV1 | |
Freshly ejaculated dog sperms were washed in PBS and extracted in 3% acetic acid, 10% glycerol, 30 mM benzaminidine. The acid extract was dialyzed against 0.2% acetic acid lyophylized and subsequently used for immunization. | |
0.1 mL | |
56 | |
Canine | |
Purified |
Western Blot, Immunocytochemistry, Immunofluorescence | |
Ds-2 | |
Western Blot, Immunocytochemistry/Immunofluorescence | |
acrosomal protein SP-10, acrosomal vesicle protein 1D11S4365, SP-10, SPACA2, sperm protein 10 | |
Mouse | |
Protein A purified | |
Primary | |
The antibody Ds-2 reacts with canine intra-acrosomal sperm protein (92 and 42 kDa), a testis-specific acrosomal vesicle protein 1 associated with the acrosomal membranes and matrix of the mature sperm. | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction