Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Intra Acrosomal Protein Antibody, mFluor Violet 610 SE, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP299874MFV610
Description
Intra Acrosomal Protein Polyclonal antibody specifically detects Intra Acrosomal Protein in Human samples. It is validated for ELISA, Immunohistochemistry (Paraffin)Specifications
Intra Acrosomal Protein | |
Polyclonal | |
ELISA, Immunohistochemistry-Paraffin | |
acrosomal protein SP-10, acrosomal vesicle protein 1D11S4365, SP-10, SPACA2, sperm protein 10 | |
Produced in rabbits immunized with purified, recombinant Human Intra Acrosomal Protein (Accession#: P26436-1; Gln22-Ile265) | |
0.1 mL | |
Primary | |
Human | |
Purified |
ELISA, Immunohistochemistry (Paraffin) | |
mFluor Violet 610 SE | |
50mM Sodium Borate | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
56 | |
Store at 4C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction