Learn More
Eisco™ Introduction to Minerals Kit
SCI_ED
Catalog No. s24662 Shop All Eisco Products
Change view
Click to view available options
Quantity:
12 Specimens
Catalog No. Quantity
S24662 12 Specimens
1 options
Catalog No. S24662 Supplier Eisco™ Supplier No. ESNG0003
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
Designed to act as gateway to wider world of mineralogy. The Introduction to Minerals Kit contains 12 specimens and specimen key.
  • For classroom and personal use
  • Added collection for first-time and experienced rock hounds
  • Average specimen size: 2 to 3 cm
  • Offers introduction into exploring earth science
Specifications
Product Type Introduction to Minerals Kit
For Use With (Application) Introduction to important minerals
Includes Hematite, magnetite, biotite, gypsum, calcite, hornblende, augite, labradorite, microcline, muscovite, olivine, quartz
Packaging Sectioned container
Quantity 12 Specimens
Specimen Size Approx. 1 in.