Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Introduction to Rocks Poster

Click to view available options
:
Laminated; Legend: Introduction to Rocks
Description
Explains that each type of rock is the result of a specific mixture of minerals subjected to a clearly defined geological process.
- Bulk of the poster is divided into three sections, one for each type of rock: igneous, sedimentary and metamorphic
- Bonus section shows and explains some of the more specialized and popular rock forms such as geodes, concretions, agates and thunder balls
- Each laminated poster includes a legend
Specifications
Specifications
Format | Poster |
Product Type | Introduction to Rocks Poster |
For Use With (Application) | Explains that each type of rock is the result of a specific mixture of minerals subjected to a clearly defined geological process |
Subject | Social Science |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction