Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
INTS1 Antibody, Alexa Fluor™ 405, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB100509AF405
Description
INTS1 Polyclonal antibody specifically detects INTS1 in Human, Mouse samples. It is validated for Western BlotSpecifications
INTS1 | |
Polyclonal | |
Western Blot | |
DKFZp586J0619, FLJ46624, integrator complex subunit 1, NET28, UNQ1821/PRO3434 | |
A synthetic peptide derived from a sequence between residues 150 and 200 of human KIAA1440 using the numbering given in entry XP_291222.2 (GeneID 26173). | |
0.1 mL | |
Primary | |
Human, Mouse | |
Purified |
Western Blot | |
Alexa Fluor 405 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
26173 | |
Store at 4C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction