Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ INTS1, Mouse, Unlabeled, Clone: 4.47,

Catalog No. MABS1984MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
MABS1984MI 100 μg Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABS1984MI Supplier MilliporeSigma™ Supplier No. MABS1984
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Integrator complex subunit 1 (UniProt Q8N201; also known as Int1) is encoded by the INTS1 (also known as KIAA1440, UNQ1821/PRO3434) gene (Gene ID 26173) in human. INTS1 is a large protein that is a component of the integrator complex that is involved in small nuclear RNAs U1 and U2 transcription and their 3&prime-box-dependent processing. During transcriptional regulation and in RNA processing, it directly binds to the C-terminal domain of the RNA polymerase II largest subunit. It has been reported that INTS1 is also involved in different cell support processes, such as the formation of a scaffold for the assembly of integrator complex. Developing INTS1 (-/-) mouse embryos show growth arrest at the early blastocyst stage and exhibit activated caspase-3 and caspase-7 that induce apoptosis within the inner cell mass of blastocyst.
TRUSTED_SUSTAINABILITY

Specifications

Antigen INTS1
Applications Western Blot
Classification Monoclonal
Clone 4.47
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Formulation Purified mouse IgG1κ in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% Sodium Azide.
Gene INTS1, KIAA1440, UNQ1821/PRO3434
Gene Accession No. Q8N201
Host Species Mouse
Immunogen GST-tagged recombinant human INTS1 internal fragment.
Purification Method Protein G Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Epigenetics & Nuclear Function
Primary or Secondary Primary
Gene ID (Entrez) NP_001073922
Test Specificity Clone 4.47 targets an epitope in the central region of human INTS1 protein.
Target Species Human
Content And Storage Stable for 1 year at 2°-8°C from date of receipt.
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.