Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ INTS1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA554488
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA554488 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA554488 Supplier Invitrogen™ Supplier No. PA554488
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LTEEEDSQTE LLIAEEKLSP EQEGQLMPRY EELAESVEEY VLDMLRDQLN RRQPIDNVSR NLLRLLTSTC GYKEVRLLAV QKLEMWLQNP K Highest antigen sequence identity to the following orthologs: Mouse - 96%, Rat - 96%.

INTS1 is a subunit of the Integrator complex, which associates with the C-terminal domain of RNA polymerase II large subunit and mediates 3-prime end processing of small nuclear RNAs U1 and U2. RNA polymerase II (Pol II) is an enzyme that is composed of twelve subunits and is responsible for the transcription of protein-coding genes. Transcription initiation requires Pol II-mediated recruitment of transcription machinery to a target promoter, thereby allowing transcription to begin.
TRUSTED_SUSTAINABILITY

Specifications

Antigen INTS1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene INTS1
Gene Accession No. Q8N201
Gene Alias 1110015K06Rik; int1; Integrator complex subunit 1; Ints1; KIAA1440; KIAA1440-like; NET28; RGD1308908; similar to KIAA1440 protein; UNQ1821/PRO3434
Gene Symbols INTS1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human INTS1. Recombinant protein control fragment (Product #RP-92855).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 26173
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.