Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ INTS10 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA589225
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA589225 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA589225 Supplier Invitrogen™ Supplier No. PA589225
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: FLTDMIIYQG QYKKAIASLH HLAALQGSIS QPQITGQGTL EHQRALIQLA TCHFALGEYR MTCEKVLDLM CYMVLPIQDG GKSQEEPSKV KPKFRKGSDL KLLPCTSKAI MPYCLHLMLA CFKLRAFTDN RDDMALGHVI VLLQQEWPRG ENLFLKAVNK ICQQGNFQYE NFFNYVTNID MLEEFAYLRT QEGGKIHLEL LPNQGMLIKH HTVTRGITKG VKEDFRLAME RQVSRCGENL MVVLHRFCIN EKILLLQTLT; Positive Samples: A-549, HL-60, HeLa, HepG2, Mouse brain; Cellular Location: Nucleus.

INTS10 is a subunit of the Integrator complex, which associates with the C-terminal domain of RNA polymerase II large subunit (POLR2A)and mediates 3-prime end processing of small nuclear RNAs U1.
TRUSTED_SUSTAINABILITY

Specifications

Antigen INTS10
Applications ELISA, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.51 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene Ints10
Gene Accession No. Q8K2A7, Q9NVR2
Gene Alias 4921521J11Rik; AI462004; C8orf35; int10; Integrator complex subunit 10; INTS10; RGD1304711
Gene Symbols Ints10
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 451-710 of human INTS10 (NP_0606122).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55174, 70885
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.