Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
INTS11 Antibody, Alexa Fluor™ 532, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB10060638AF532
Description
INTS11 Polyclonal antibody specifically detects INTS11 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
INTS11 | |
Polyclonal | |
Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin | |
cleavage and polyadenylation specific factor 3-like, Cleavage and polyadenylation-specific factor 3-like protein, CPSF3-like protein, EC 3.1.27, EC 3.1.27.-, FLJ20542, INT11, integrator complex subunit 11, INTS11FLJ13294, Protein related to CPSF subunits of 68 kDa, RC68, RC-68CPSF73L, related to CPSF subunits 68 kDa | |
The immunogen recognized by this antibody maps to a region between residue 550 and 600 of human integrator complex subunit 11 using the numbering given in entry NP_060341.2 (GeneID 54973). | |
0.1 mL | |
Primary | |
Human, Mouse | |
Purified |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Alexa Fluor 532 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
54973 | |
Store at 4C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction