Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
INTS11 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB10060638
Description
INTS11 Polyclonal specifically detects INTS11 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin.Specifications
INTS11 | |
Polyclonal | |
Unconjugated | |
Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.09% Sodium Azide | |
cleavage and polyadenylation specific factor 3-like, Cleavage and polyadenylation-specific factor 3-like protein, CPSF3-like protein, EC 3.1.27, EC 3.1.27.-, FLJ20542, INT11, integrator complex subunit 11, INTS11FLJ13294, Protein related to CPSF subunits of 68 kDa, RC68, RC-68CPSF73L, related to CPSF subunits 68 kDa | |
Rabbit | |
68 kDa | |
100 μL | |
Primary | |
Human, Mouse. Based on 100% sequence identity, this antibody is predicted to react with Horse, Panda, Orangutan, Rhesus Monkey, Gorilla, Chimpanzee, White-tufted-ear marmoset, Crab-eating macaque, African elephant and Gray short-tailed opossum. | |
Store at 4C. Do not freeze. |
Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
1.0 mg/mL | |
Western Blot 1:2000-1:10000, Immunohistochemistry 1:1000 - 1:5000, Immunoprecipitation 2-10 μg/mg lysate, Immunohistochemistry-Paraffin 1:1000 - 1:5000 | |
Q5TA45 | |
CPSF3L | |
The immunogen recognized by this antibody maps to a region between residue 550 and 600 of human integrator complex subunit 11 using the numbering given in entry NP_060341.2 (GeneID 54973). | |
Affinity Purified | |
RUO | |
54973 | |
Human, Mouse | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction