Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
INTS11 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$382.00 - $646.00
Specifications
Antigen | INTS11 |
---|---|
Dilution | Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin 1:50 - 1:200 |
Applications | Immunohistochemistry, Immunohistochemistry (Paraffin), KnockDown |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
INTS11 Polyclonal antibody specifically detects INTS11 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), Knockdown.Specifications
INTS11 | |
Immunohistochemistry, Immunohistochemistry (Paraffin), KnockDown | |
Unconjugated | |
RUO | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
54973 | |
This antibody was developed against Recombinant Protein corresponding to amino acids:LVGQAEPESVLLVHGEAKKMEFLKQKIEQELRVNCYMPANGETVTLPTSPSIPVGISLGLLKREMAQGLLPEAKKPRLLHGTLIMKDSNFRLVSSEQALK | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
Polyclonal | |
Rabbit | |
Human, Mouse, Rat | |
cleavage and polyadenylation specific factor 3-like, Cleavage and polyadenylation-specific factor 3-like protein, CPSF3-like protein, EC 3.1.27, EC 3.1.27.-, FLJ20542, INT11, integrator complex subunit 11, INTS11FLJ13294, Protein related to CPSF subunits of 68 kDa, RC68, RC-68CPSF73L, related to CPSF subunits 68 kDa | |
CPSF3L | |
IgG | |
Affinity Purified | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title