Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ INTS2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA561998
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA561998 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA561998 Supplier Invitrogen™ Supplier No. PA561998
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: ICLPTEEEKA NGVNPDSLLR NVQSVITTSA PNKGMEEGED NLLCNLREVQ CLICCLLHQM YIADPNIAKL VHFQGYPCEL L Highest antigen sequence identity to the following orthologs: Mouse - 84%, Rat - 83%.

INTS2 is a subunit of the Integrator complex, which associates with the C-terminal domain of RNA polymerase II large subunit (POLR2A; MIM 180660) and mediates 3-prime end processing of small nuclear RNAs U1 (RNU1; MIM 180680) and U2 (RNU2; MIM 180690) (Baillat et al., 2005 [PubMed 16239144]).[supplied by OMIM, Mar 2008].
TRUSTED_SUSTAINABILITY

Specifications

Antigen INTS2
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene INTS2
Gene Accession No. Q9H0H0
Gene Alias 2810417D08Rik; AA408260; AI987735; int2; Integrator complex subunit 2; INTS2; KIAA1287; mKIAA1287; RGD1305547
Gene Symbols INTS2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human INTS2. Recombinant protein control fragment (Product #RP-101170).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57508
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.