Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
INTS5 Antibody, DyLight 488, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB10060461G
Description
INTS5 Polyclonal antibody specifically detects INTS5 in Human samples. It is validated for Western BlotSpecifications
INTS5 | |
Polyclonal | |
Western Blot | |
Int5, integrator complex subunit 5, KIAA1698INT5 | |
The immunogen recognized by this antibody maps to a region between residue 975 and the C-terminus of human integrator complex subunit 5 using the numbering given in entry NP_085131.1 (GeneID 80789). | |
0.1 mL | |
Primary | |
Human | |
Purified |
Western Blot | |
DyLight 488 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
80789 | |
Store at 4C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction