Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ INTS5 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5109865
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5109865 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5109865 Supplier Invitrogen™ Supplier No. PA5109865
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: DVVQEVQQVL SEFIRANPKA WAPVISAWSI DLMGQLSSTY SGQHQRVPHA TGALNELLQL WMGCRATRTL MDIYVQCLSA LIGSCPDACV DALLDTSVQH SPHFDWVVAH IGSSFPGTII SRVLSCGLKD FCVHGGAGGG AGSSGGSSSQ TPSTDPFPGS PAIPAEKRVP KIASVVGILG HLASRHGDSI RRELLRMFHD SLAGGSGGRS GDPSLQATVP FLLQLAVMSP ALLGTVSGEL VDCLKPPAVL SQLQQHLQGF PREELDNMLN LAVHLVSQAS G.

INTS5 is a subunit of the Integrator complex, which associates with the C-terminal domain of RNA polymerase II large subunit.
TRUSTED_SUSTAINABILITY

Specifications

Antigen INTS5
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.97 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene INTS5
Gene Accession No. Q6P9B9, Q8CHT3
Gene Alias 1110055N21Rik; INT5; integrator complex subunit 5; Ints5; KIAA1698; mKIAA1698; RGD1309370
Gene Symbols INTS5
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 120-400 of human INTS5 (NP_085131.1).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 109077, 80789
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.