Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ INTS6 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA551643
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA551643 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA551643 Supplier Invitrogen™ Supplier No. PA551643
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: VVNNHIGGKG PPAPTTQAQP DLIKPLPLHK ISETTNDSII HDVVENHVAD QLSSDITPNA MDTEFSASSP ASLLERPTNH MEALGHDHLG TNDLTVGGFL ENHEEPRDKE QCAEENIPAS SLNKGKKLMH CRSHEEVNTE LKAQ Highest antigen sequence identity to the following orthologs: Mouse - 85%, Rat - 83%.

DEAD box proteins, characterized by the conserved motif Asp-Glu-Ala-Asp (DEAD), are putative RNA helicases. INTS6 is a DEAD box protein that is part of a complex that interacts with the C-terminus of RNA polymerase II and is involved in 3' end processing of snRNAs. In addition, this gene is a candidate tumor suppressor and located in the critical region of loss of heterozygosity (LOH). DEAD box proteins, characterized by the conserved motif Asp-Glu-Ala-Asp (DEAD), are putative RNA helicases. The protein encoded by this gene is a DEAD box protein that is part of a complex that interacts with the C-terminus of RNA polymerase II and is involved in 3' end processing of snRNAs. In addition, this gene is a candidate tumor suppressor and located in the critical region of loss of heterozygosity (LOH). Three transcript variants encoding two different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen INTS6
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene INTS6
Gene Accession No. Q9UL03
Gene Alias 2900075H24Rik; AI480962; Dbi1; DBI-1; Ddx26; DDX26A; DEAD box protein; DEAD/H (Asp-Glu-Ala-Asp/His) box polypeptide 26; DICE1; DKFZP434B105; HDB; INT6; integrator complex subunit 6; INTS6; LRRGT00024; Notch2l; Notchl2; Protein DDX26; Protein deleted in cancer 1; RNA helicase HDB
Gene Symbols INTS6
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human INTS6. Recombinant protein control fragment (Product #RP-95936).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 26512
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.