Learn More
Invitrogen™ INTS8 Polyclonal Antibody

Description
Positive test controls include: Mouse brain. Immunogen sequence: VTEFSPKLRQ VMLNEMLLLD IHTHEAGTGQ AGERPPSDLI SRVRGYLEMR LPDIPLRQVI AEECVAFMLN WRENEYLTLQ VPAFLLQSNP YVKLGQLLAA TCKELPGPKE SRRTAKDLWE VVVQICSVSS QHKRGNDGRV SLIKQRESTL GIMYRSELLS FIKKLREPLV LTIILSLFVK LHNVREDIVN DITAEHISIW P.
Specifications
Specifications
| Antigen | INTS8 |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Concentration | 0.62 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.05% ProClin 300; pH 7.3 |
| Gene | INTS8 |
| Gene Accession No. | Q75QN2, Q80V86 |
| Gene Alias | C8orf52; FLJ20530; Int8; integrator complex subunit 8; INTS8; MGC131633; Protein kaonashi-1 |
| Gene Symbols | INTS8 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.