Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
INTS8 Rabbit anti-Human, Mouse, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
$173.05 - $405.08
Specifications
| Antigen | INTS8 |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Form | Purified |
Description
INTS8 Polyclonal antibody specifically detects INTS8 in Human, Mouse samples. It is validated for Western BlotSpecifications
| INTS8 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse | |
| C8orf52, FLJ20530, INT8, integrator complex subunit 8, protein kaonashi-1 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 600-800 of human INTS8 (NP_060334.2). | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Cell Biology | |
| PBS, 50% glycerol, pH7.3 | |
| 55656 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title