Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Inulin

Catalog No. p-7024162
Click to view available options
Quantity:
10 g
50 g
250 g

Catalog No. AAA1842509

Supplier: Thermo Scientific Chemicals A1842509

Only null left
Add to Cart

CAS: 9005-80-5

Mixture of fructose polymers that serves as carbohydrate storage in plantsInulin is used in the treatment of high blood fats, cholesterol and triglycerides. It is also used for weight loss, constipation and also serves as a food additive to improve taste and as a vaccine adjuvant. Further, it is utilized to replace sugar, fat and flour. It is also used to measure kidney function by determining the glomerular filtration rate (GFR). In addition to this, it is used to replace some of the flour in baked goods.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Mixture of fructose polymers that serves as carbohydrate storage in plantsInulin is used in the treatment of high blood fats, cholesterol and triglycerides. It is also used for weight loss, constipation and also serves as a food additive to improve taste and as a vaccine adjuvant. Further, it is utilized to replace sugar, fat and flour. It is also used to measure kidney function by determining the glomerular filtration rate (GFR). In addition to this, it is used to replace some of the flour in baked goods.

Solubility
Soluble in water.

Notes
Yields D-fructose and D-glucose after acid hydrolysis.Hygroscopic. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 9005-80-5
MDL Number MFCD00131407

Specifications

Odor Odorless
Assay Percent Range 0.98
Quantity 10 g
Merck Index 14,5004
Solubility Information Soluble in water.
Chemical Name or Material Inulin
EINECSNumber 232-684-3
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.