Alfa Aesar™ Inulin

Manufacturer: Alfa Aesar™ A1842530

Quantity 250g

 View more versions of this product

Catalog No. AAA1842530

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 9005-80-5
    MDL Number MFCD00131407
    Merck Index 14,5004
    Chemical Name or Material Inulin

    Specifications

    Assay Percent Range 0.98
    TSCA TSCA
    MDL Number MFCD00131407
    CAS 9005-80-5
    Merck Index 14,5004
    Quantity 250g
    Odor Odorless
    Chemical Name or Material Inulin

    Description

    Applications
    Mixture of fructose polymers that serves as carbohydrate storage in plantsInulin is used in the treatment of high blood fats, cholesterol and triglycerides. It is also used for weight loss, constipation and also serves as a food additive to improve taste and as a vaccine adjuvant. Further, it is utilized to replace sugar, fat and flour. It is also used to measure kidney function by determining the glomerular filtration rate (GFR). In addition to this, it is used to replace some of the flour in baked goods.

    Solubility
    Soluble in water.

    Notes
    Yields D-fructose and D-glucose after acid hydrolysis.Hygroscopic. Incompatible with strong oxidizing agents.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit