Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invariant NK T Cell Mouse anti-Human, Brilliant Violet 605, Clone: 6B11, BD Optibuild™

Catalog No. BDB743999 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB743999 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB743999 Supplier BD Biosciences Supplier No. 743999
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Monoclonal antibody 6B11 reacts with a unique determinant in the CDR3 region of the invariant (Vα24-JαQ) TCR chain. Thus 6B11 identifies a subset of all Vα24 positive T cells. The binding of the PE-conjugate of 6B11 can be partially blocked by other antibodies specific for Vα24 (BD Biosciences Pharmingen, data not shown). This invariant chain is expressed on invariant NK T cells, which are a small subset of T lymphocytes that also express other NK cell molecules such as CD161. Invariant NK T cells have been reported to play an immunoregulatory role. The Vα24-JαQ interacts with the CD1d protein. Invariant NK T cells can be specifically activated by CD1d-presented antigen resulting in rapid production of IL-4 and IFN (also known as IFN-γ). Studies of NK T cells describe their role in anti-tumor immunity and autoimmunity.

This antibody is conjugated to BD Horizon™ BV605 which is part of the BD Horizon Brilliant™ Violet family of dyes. With an Ex Max of 407nm and Em Max of 602nm, BD Horizon BV605 can be excited by a violet laser and detected with a standard 610/20nm filter set. BD Horizon BV605 is a tandem fluorochrome of BD Horizon BV421 and an acceptor dye with an Em max at 605nm. Due to the excitation of the acceptor dye by the green (532 nm) and yellow-green (561 nm) lasers, there will be significant spillover into the PE and BD Horizon PE-CF594 detectors off the green or yellow-green lasers. BD Horizon BV605 conjugates are very bright, often exhibiting brightness equivalent to PE conjugates and can be used as a third color off of the violet laser. BV605 is a tandem fluorochrome that combines BD Horizon BV421 and Cy™3.5. These conjugates are very bright, exhibiting similar brightness to equivalent PE conjugates. Due to the excitation of Cy3.5 by the green (532nm) and yellow-green (561nm) lasers, there will be significant spillover of the BD Horizon BV605 signal into the PE and BD Horizon PE-CF594 detectors off the green or yellow-green lasers.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Invariant NK T Cell
Classification Monoclonal
Clone 6B11
Concentration 0.2mg/mL
Conjugate Brilliant Violet 605
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Gene Alias Vα24JαQ TCR Chain
Host Species Mouse
Immunogen Peptide from CDR3 loop of the invariant TCR V24 sequence
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.