Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Inversin Rabbit anti-Human, Mouse, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP303537100UL
Description
Inversin Polyclonal specifically detects Inversin in Human, Mouse samples. It is validated for Western Blot.Specifications
| Inversin | |
| Polyclonal | |
| PBS with 50% glycerol, pH7.3. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 27130 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| inversin, Inversion of embryo turning homolog, INVMGC133081, KIAA0573, MGC133080, nephrocystin 2, nephrocystin-2, nephronophthisis 2 (infantile), NPH2, NPHP2inversion of embryonic turning | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-90 of human Inversin (NP_055240.2). | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction