Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ siPORT™ NeoFX™ Transfection Agent with manual

Catalog No. p-4924024
Encompass_Preferred
Click to view available options
Quantity:
400 μL
1 mL
Product Type:
Transfection Reagent
Transfection Reagent with Manual

Includes

One tube containing 0.4mL

Lipid-based formulation can be used to efficiently transfect adherent cells with siRNA as they are subcultured—without increased cytotoxicity

  • Versatile-works with a broad range of cell lines
  • Reproducible-produces consistent results, lot-to-lot, plate-to-plate, and well-to-well
  • Efficient-performs with high siRNA transfection efficiency, allowing use of low siRNA concentrations to minimize nonspecific effects
  • Fast-reverse-transfects cells as they are plated, saving a day
  • Reverse-Transfect Cells as They are Plated, Saving a Day
  • Optimized for Lower siRNA Concentrations and High Reproducibility
  • Compatible with a Broad Range of Cell Lines

Cell Culture, RNAi Transfection, Synthetic siRNA Transfection, Transfection

Order Info

Shipping Condition: Room Temperature

TRUSTED_SUSTAINABILITY

Specifications

Classification Animal Origin-Free
Content And Storage Store at 4°C.
Cell Type Stem Cells, Primary Cells, Mammalian
Format 6-well Plate, 12-well Plate, 24-well Plate, 48-well Plate, 96-well Plate, Flasks
Sample Type Synthetic siRNA
Transfection Technique Lipid-based Transfection
For Use With (Application) Transfection
High-throughput Compatibility High-throughput Compatible
Product Line Ambion™, siPORT™
Product Type Transfection Reagent
Quantity 400 μL
Serum Compatible Yes
Shipping Condition Room Temperature
Show More Show Less
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.