Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ BioPrime™ Total Genomic Labeling System

Catalog No. 18097011
Encompass_Preferred
Click to view available options
Quantity:
10 Reactions
30 Reactions

Catalog No. 18-097-011

Supplier: Invitrogen™ 18097011

Only null left
Add to Cart
Add to Cart

Complete genomic DNA labeling kits designed for use in array comparative genomic hybridization applications (aCGH) that allows for better call rates with precious samples

These complete, all-inclusive kits are optimized to work across the widest range of sample input material with no need for pre-amplification. Highly concentrated exo-Klenow fragment of DNA polymerase I in a component limited reaction allow for consistent, robust DNA yields of ∽8μg with as little as 50ng of genomic DNA input. An optimized dye labeled nucleotide mix with new dNTP linkers, novel, application-specific Alexa Fluor 3 and 5 dyes, and improved buffer chemistry reduce labeling variation and increase signal to noise on arrays. Excitation and emission spectra of Alexa Fluor 3 and 5 dyes are suited for conventional two color scanners with no need to change settings. Complete kits are batch tested to assure quality in performance from lot to lot.
Contents and Storage:
The simplified kits contains Alexa Fluor 3 and Alexa Fluor 5 2x reaction mix, Exo-Klenow fragment (40U/μL), 5mM EDTA, TE Buffer, and control DNA (Salmon Sperm). The BioPrime Total Genomic Labeling System also contains PurelinkTM Genomic DNA purification columns and purification buffers (the BioPrime Total Genomic Labeling Module does not). New master mix formulation streamlines reaction set-ups, increasing consistency.

  • Higher signal to noise with Alexa Fluor™ 3 and 5 dyes
  • Less channel bias due to novel reaction formulation
  • Widest range of input material (50ng - 3μg)
  • Simplified workflow with master mix formulation

Array CGH, ChIP-on-Chip, Chromatin Biology, DNA and RNA Purification and Analysis, DNA Labeling, Gene Expression Analysis and Genotyping, Genotyping and Genomic Profiling, Nucleic Acid Labeling and Oligo Synthesis, RNAi, Epigenetics and Non-Coding RNA Research

TRUSTED_SUSTAINABILITY

Specifications

Detection Method Fluorescence
Label or Dye Alexa Fluor 3, Alexa Fluor 5
Final Product Type Probes (Labeled DNA)
Labeling Target Genomic DNA
Includes Label or Dye Yes
Labeling Method Direct Labeling
Product Line BioPrime
Product Type Total Genomic Labeling System
Quantity 30 Reactions

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.