Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ChargeSwitch™ gDNA Plant Kit
GREENER_CHOICE

Catalog No. p-4926407
Encompass_Preferred
Change view
Click to view available options
No. of Reactions:
96 Preps
960 Preps
Sample Type:
Fungi, Plants
Leaf, Seeds
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. No. of Reactions Sample Type
CS18000 96 Preps Fungi, Plants
CS1800010 960 Preps Leaf, Seeds
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. CS18000 Supplier Invitrogen™ Supplier No. CS18000
Only null left
Add to Cart
Add to Cart

Includes

ChargeSwitch Magnetic Beads (25 mg/mL in 10 mM MES, pH 5.0, 10 mM NaCl, 0.1% Tween 20)
ChargeSwitch Lysis Buffer (L18)
ChargeSwitch Precipitation Buffer (N5)
ChargeSwitch Wash Buffer (W12)
ChargeSwitch Elution Buffer (E6; 10 mM Tris-HCl, pH 8.5 with 1 mM EDTA)
RNase A (5 mg/mL in 10 mM Tris-HCl, pH 8.5, 10 mM EDTA)
10% SDS in deionized water
10% Detergent (D1)

The ChargeSwitch™ gDNA Plant Kit uses magnetic bead-based technology to allow rapid and efficient isolation of genomic DNA from fungal and plant samples, including leaves, tissues and seeds, in less than 15 minutes after lysate preparation.

The ChargeSwitch™ gDNA Plant Kit uses magnetic bead-based technology to allow rapid and efficient isolation of genomic DNA from fungal and plant samples, including leaves, tissues and seeds, in less than 15 minutes after lysate preparation.

• Optimized for higher-purity DNA extraction
• Compatible with high-throughput protocols
• Ideal for a variety of downstream applications
• Less hazardous, less waste

Order Info

Shipping Condition: Room Temperature

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage

All components of the ChargeSwitch™ gDNA Plant Kit are shipped at room temperature. Upon receipt, store RNase A at 4°C. Store all other kit components at room temperature.

Components:
• ChargeSwitch™ Magnetic Beads (25 mg/mL in 10 mM MES, pH 5.0, 10 mM NaCl, 0.1% Tween 20)
• ChargeSwitch™ Lysis Buffer (L18)
• ChargeSwitch™ Precipitation Buffer (N5)
• ChargeSwitch™ Wash Buffer (W12)
• ChargeSwitch™ Elution Buffer (E6; 10 mM Tris-HCl, pH 8.5 with 1 mM EDTA)
• RNase A (5 mg/mL in 10 mM Tris-HCl, pH 8.5, 10 mM EDTA)
• 10% SDS in deionized water
• 10% Detergent (D1)

Format Kit
Isolation Technology ChargeSwitch Chemistry
Sample Type Fungi, Plants
For Use With (Application) PCR, Southern Blotting, Sequencing, Cloning
High-throughput Compatibility High-throughput Compatible
No. of Reactions 96 Preps
Product Line ChargeSwitch
Product Type gDNA Plant Kit
Quantity 96 Preps
Shipping Condition Room Temperature
Target Genomic DNA
Test Time 15 min.
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.