Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DNA Polymerase I

DNA polymerase with 5'→3' and 3'→5' exodeoxyribonuclease activities

Supplier:  Invitrogen™ 18010017

Encompass_Preferred

 View more versions of this product

Catalog No. 18-010-017


Only null left
Edge
Add to Cart

Includes: 1000 units of DNA polymerase in a storage buffer of 50mM potassium phosphate (pH 7.0), 100mM KCl, 1mM DTT, and 50% (v/v) glycerol. It is supplied at 5-10 units/μL.

Description

Description

Performance and quality testing
Endodeoxyribonuclease assay; efficiency of DNase I-dependent nick translation determined.

Unit definition

One unit incorporates 10nMol of total deoxyribonucleotide into acid-precipitable material in 30 min at 37°C using template/primer.

Unit reaction conditions

50mM potassium phosphate (pH 7.0), 6.7mM MgCl2 , 1mM 2-mercaptoethanol, 80μg/mL template/primer, 32μM dTTP, 69nM [3H]dTTP, and enzyme in 100μL for 30 min at 37°C. Also incorporates biotinylated nucleotides.

  • Applications: DNase I-dependent nick translation, second-strand synthesis in cDNA cloning, fill-in of 5' overhangs
  • Source: purified from E. coli expressing the DNA Polymerase I gene on a plasmid

Cloning, Restriction Enzyme Cloning

Order Info

Shipping Condition: Wet Ice

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Store in freezer (-5 to -30°C).
Liquid
3' - 5', 5' - 3'
Wet Ice
DNA Polymerase
DNA Polymerase I
250 U
No
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.

For Research Use Only. Not for use in diagnostic procedures.